Learn More
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
5488.73 DKK - 13196.95 DKK
Product Code | Brand | Holds | For Use With (Equipment) | No. of Racks | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | For Use With (Equipment) | No. of Racks | Price | Quantity & Availability | ||||
12908344
|
Thermo Scientific™
RSK13SD4 |
Boxes (48/4 Racks) | Forma 900, Forma 7000, TSE and TSD Series (13 cu.ft.) | 4 |
5773.66 DKK
Each |
In Stock
Log in to see stock. |
||||
11329351
|
Thermo Scientific™
RSK13SD5 |
Boxes (48/4 Racks) | HERAfreeze HFU B Series, Revco ExF and DxF Series (13 cu.ft.) | 4 |
5488.73 DKK
Each |
Estimated Shipment: 31-10-2025
Log in to see stock. |
||||
12918344
|
Thermo Scientific™
RSK17SD4 |
Boxes (80/4 Racks) | Forma 900, Forma 7000, TSE and TSD Series (17 cu.ft.) | 4 |
6778.43 DKK
Each |
Estimated Shipment: 14-11-2025
Log in to see stock. |
||||
11339351
|
Thermo Scientific™
RSK17SD5 |
Boxes (80/4 Racks) | HERAfreeze HFU B Series, Revco ExF and DxF Series (17 cu.ft.) | 4 |
6770.94 DKK
Each |
Estimated Shipment: 13-11-2025
Log in to see stock. |
||||
12928344
|
Thermo Scientific™
RSK23SD4 |
Boxes (100/5 Racks) | Forma 900, Forma 7000, TSE and TSD Series (23 cu.ft.) | 5 |
8548.02 DKK
Each |
Estimated Shipment: 31-10-2025
Log in to see stock. |
||||
11349351
|
Thermo Scientific™
RSK23SD5 |
Boxes (80/5 Racks) | HERAfreeze HFU B Series, Revco ExF and DxF Series | 5 |
8548.02 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
||||
11810543
|
Thermo Scientific™
RSK300SD4 |
Boxes (75/3 Racks) | Thermo Scientific TDE Series (14.9 cu.ft.) | 3 |
5646.19 DKK
Each |
Estimated Shipment: 28-10-2025
Log in to see stock. |
||||
11359351
|
Thermo Scientific™
RSK300SD5 |
Boxes (60/3 Racks) | Thermo Scientific TDE Series (14.9 cu.ft.) | 3 |
5616.20 DKK
Each |
In Stock
Log in to see stock. |
||||
11820543
|
Thermo Scientific™
RSK400SD4 |
Boxes (100/4 Racks) | Thermo Scientific TSX and TDE Series (19 cu.ft.) | 4 |
7498.26 DKK
Each |
In Stock
Log in to see stock. |
||||
11369351
|
Thermo Scientific™
RSK400SD5 |
Boxes (80/4 Racks) | Thermo Scientific TSX and TDE Series (19 cu.ft.) | 4 |
7468.28 DKK
Each |
In Stock
Log in to see stock. |
||||
11830543
|
Thermo Scientific™
RSK500SD4 |
Boxes (125/5 Racks) | Thermo Scientific TSX and TDE Series (24.1 cu.ft.) | 5 |
9447.81 DKK
Each |
In Stock
Log in to see stock. |
||||
11379351
|
Thermo Scientific™
RSK500SD5 |
Boxes (100/5 Racks) | Thermo Scientific TSX and TDE Series (24.1 cu.ft.) | 5 |
9372.83 DKK
Each |
Estimated Shipment: 31-10-2025
Log in to see stock. |
||||
11840543
|
Thermo Scientific™
RSK600SD4 |
Boxes (150/6 Racks) | Thermo Scientific TSX and TDE Series (28.8 cu.ft.) | 6 |
11322.38 DKK
Each |
In Stock
Log in to see stock. |
||||
17708049
|
Thermo Scientific™
RSK600SD5 |
Boxes (120/6 Racks) | Thermo Scientific TSX and TDE Series (28.8 cu.ft.) | 6 |
11193.95 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11850543
|
Thermo Scientific™
RSK700SD4 |
Boxes (175/7 Racks) | Thermo Scientific TSX Series (33.5 cu.ft.) | 7 |
13196.95 DKK
Each |
Estimated Shipment: 31-10-2025
Log in to see stock. |
||||
11399351
|
Thermo Scientific™
RSK700SD5 |
Boxes (140/7 Racks) | Thermo Scientific TSX Series (33.5 cu.ft.) | 7 |
13121.96 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
||||
Description
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features:
- Stainless steel racks
- Boxes
- Cell dividers
Warranty varies by country. Please contact us for details.
Specifications
100 |