missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium Chloride, ACS Reagent Grade, Scharlab
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium Chloride, ACS Reagent Grade, Scharlab
Product Code. 113285445 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable.
View return policy
Product Code. 11328544
Brand: Scharlab SO0227005P
This item is not returnable.
View return policy
Description
- Synonyms: Salt, Common salt, Rock salt, Sea salt
- Molecular Formula: NaCl
- Molar Mass: 58.44g/mol
- CAS: 7647-14-5
- EIN ECS No.: 231-598-3
Specifications
Chemical Identifiers
CAS | 7647-14-5 |
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
IUPAC Name | sodium chloride |
SMILES | [Na+].[Cl-] |
Specifications
Boiling Point | 1461°C |
Melting Point | 801°C |
pH | 5.0–8.0 (5%, water) |
Quantity | 5 kg |
Formula Weight | 58.44g/mol |
Percent Purity | 99.5% min (argentometric) |
Grade | ACS Reagent |
Identification | Passes Test |
Loss on Drying | 0.5% max (105°C, 2h) |
Vapor Pressure | 1.3 hPa (at 865°C) |
Product Suggestions
You may be interested in these alternatives
-
Product Code. 10735921
-
-
-
Product Code. 15621310
-
Product Code. 15631310
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 6
-
Catalog No. 10703811
kr 1.480,00 / 500g -
Catalog No. 10793611
kr 525,00 / 1kg -
Catalog No. 10117113
kr 283,43 / 100g -
Catalog No. 10112640
kr 177,00 / 1kg -
Catalog No. 10092740
kr 224,95 / 1kg -
Catalog No. 10401841
kr 239,41 / 500mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Chloride, ACS Reagent Grade, Scharlab >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction