Learn More
Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate, Thermo Scientific Chemicals
181.46 DKK - 1739.59 DKK
Chemical Identifiers
CAS | 7681-38-1 |
---|---|
Molecular Formula | HNaO4S |
Molecular Weight (g/mol) | 120.05 |
MDL Number | MFCD00003531 |
InChI Key | WBHQBSYUUJJSRZ-UHFFFAOYSA-M |
Synonym | sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect |
PubChem CID | 516919 |
SMILES | [Na+].OS([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11385045
|
Thermo Scientific Alfa Aesar
B25587.30 |
250 g |
181.46 DKK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11375045
|
Thermo Scientific Alfa Aesar
B25587.0B |
1000 g |
500.88 DKK
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15405095
|
Thermo Scientific Alfa Aesar
B25587.0I |
5000 g |
1739.59 DKK
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7681-38-1 | |
120.05 | |
WBHQBSYUUJJSRZ-UHFFFAOYSA-M | |
516919 |
HNaO4S | |
MFCD00003531 | |
sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect | |
[Na+].OS([O-])(=O)=O |
Specifications
177°C to 180°C | |
1.0 | |
MFCD00003531 | |
14,8586 | |
WBHQBSYUUJJSRZ-UHFFFAOYSA-M | |
sodium;hydrogen sulfate | |
516919 | |
≥90% | |
Hygroscopic | |
remainder mainly sodium sulfate |
7681-38-1 | |
HNaO4S | |
UN3260 | |
sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect | |
[Na+].OS([O-])(=O)=O | |
120.05 | |
120.06 | |
Odorless | |
2.435 | |
Sodium hydrogen sulfate, anhydrous |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 231-665-7
missing translation for 'rtecsNumber' : VZ1860000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydrogen sulfate, anhydrous
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.