Learn More
Sofosbuvir impurity H, MedChemExpress
MedChemExpress Sofosbuvir impurity H, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Brand: Medchem Express HY-I0938
This item is not returnable.
View return policy
Description
Sofosbuvir impurity H, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.Specifications
Sofosbuvir impurity H | |
Research | |
5 mg | |
633.56 | |
Research | |
COVID-19-anti-virus |
Off-White | |
C29H33FN3O10P | |
O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](OC(C2=CC=CC=C2)=O)[C@](F)(C)[C@H](N3C=CC(NC3=O)=O)O1)OC4=CC=CC=C4 | |
633.56 | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.