Learn More
Sofosbuvir impurity H, MedChemExpress
MedChemExpress Sofosbuvir impurity H, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
17825.63 DKK - 41589.79 DKK
Specifications
Chemical Name or Material | Sofosbuvir impurity H |
---|---|
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C29H33FN3O10P |
SMILES | O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](OC(C2=CC=CC=C2)=O)[C@](F)(C)[C@H](N3C=CC(NC3=O)=O)O1)OC4=CC=CC=C4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16472469
|
Medchem Express
HY-I0938 |
1 mg |
17825.63 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16411258
|
Medchem Express
HY-I0938 |
5 mg |
41589.79 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sofosbuvir impurity H, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.Specifications
Sofosbuvir impurity H | |
Research | |
O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](OC(C2=CC=CC=C2)=O)[C@](F)(C)[C@H](N3C=CC(NC3=O)=O)O1)OC4=CC=CC=C4 | |
633.56 | |
Solid |
Off-White | |
C29H33FN3O10P | |
633.56 | |
Research | |
COVID-19-anti-virus |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.