Learn More
Memmert™ Software Atmocontrol, Base Licence For 1 Unit, Iq/Oq Doc. Available In De And En, For S450
Memmert™ AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Brand: Memmert™ S450FDAQ1
Description
- Software conforming to FDA AtmoCONTROL
- Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
- Base licence for the control of one unit
- Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA) | |
Models U, UF, TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 106 | |
Software |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Memmert AtmoCONTROL (EN)
-
Memmert ControlCOCKPIT (EN)
Your input is important to us. Please complete this form to provide feedback related to the content on this product.