missing translation for 'onlineSavingsMsg'
Learn More

Solveco SolvClear Xylene Substitute

Xylene substitute for deparaffinization of tissue samples.

Brand:  Solveco 1651


Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 18022193

  • 1160.00 DKK / 15 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

SolvClear is an advanced xylene substitute specifically designed for the deparaffinization of tissue samples in histological and pathological laboratories. Unlike traditional aromatic solvents like xylene, SolvClear offers a safer and more environmentally conscious alternative.

The product is engineered to be less toxic and less harmful to both laboratory personnel and the environment, reducing exposure to potentially hazardous fumes and decreasing the ecological footprint associated with chemical disposal. SolvClear maintains the effectiveness of xylene in dissolving paraffin wax, ensuring high-quality sample preparation while promoting a healthier and more sustainable laboratory environment.

Specifications

Specifications

Liquid
Clear Colorless
Histological
90622-57-4
5 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Solveco SolvClear Xylene Substitute > 5 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.