Learn More
Eppendorf™ Spare Sealing
For use with Eppendorf Rotors
Brand: Eppendorf™ 5820705000
790.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spare sealing for rotor lids
- Available for a wide range of products
Specifications
Spare Sealing | |
Rotor A-2-DWP-AT | |
Pack of 2 |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Eppendorf™ Spare Sealing > Aerosol Tight Cap 400mL
Spot an opportunity for improvement?Share a Content Correction