Learn More
Invitrogen™ Custom DNA Oligos Synthesis Services
Custom Primer
Brand: Invitrogen™ 10668051
24.38 DKK valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10ÎĽmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
All Scales, 25 nmol | |
Custom Conc Standard Primer, 25nmol | |
Custom Conc Standard Primer |