Learn More
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10145500
|
KGW Isotherm™
1292 |
3 L |
4495.00 DKK
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
10507901
|
KGW Isotherm™
1293 |
5 L |
5240.00 DKK
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
Description
- Low evaporation rate
- Good tipping stability
- Fully silvered as standard
- Brushed aluminum cover
- Includes handle and loose lied lid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KGW Isotherm™ Spherical Dewar Flasks