Learn More
Fritsch™ Stainless Steel 1.4404/316L Sieve Ring
Brand: Fritsch™ 44.1010.10
This item is not returnable.
View return policy
Specifications
PULVERISETTE 14 Variable Speed Rotor Mill | |
Stainless Steel (316L) | |
0.12 mm | |
Trapezoid holes |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.