Learn More
SI Analytics™ Stainless Steel Brackets
Protect your viscometer from breakage. SI Analytics™ Stainless Steel Brackets are designed to ensure that the viscometers are held in a vertical position.
Brand: SI Analytics™ 285405019
1880.16 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- The maximum deviation is < 1°
- In application in conjunction with SCHOTT Instruments and other commercially available see-through thermostats the viscometers can only be used with the appropriate stand or bracket
- Provide protection for viscometers
- Allow viscometers to be held vertically
Specifications
Bracket | |
Reverse Flow Viscometer | |
Stainless Steel | |
For manual and automatic measurements |
Product Content Correction
Product Title
SI Analytics™ Stainless Steel Brackets > For Use With: For All Reverse flow Viscometer
Spot an opportunity for improvement?Share a Content Correction