Learn More
SI Analytics™ Stainless Steel Brackets
Protect your viscometer from breakage. SI Analytics™ Stainless Steel Brackets are designed to ensure that the viscometers are held in a vertical position.
1540.00 DKK - 2120.00 DKK
Specifications
Product Type | Bracket |
---|---|
Material | Stainless Steel |
For Use With (Application) | For manual and automatic measurements |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
10751201
|
SI Analytics™
285405043 |
Ubbelohde Viscometer |
1540.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
10241881
|
SI Analytics™
285405019 |
Reverse Flow Viscometer |
2120.00 DKK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
Description
- The maximum deviation is < 1°
- In application in conjunction with SCHOTT Instruments and other commercially available see-through thermostats the viscometers can only be used with the appropriate stand or bracket
- Provide protection for viscometers
- Allow viscometers to be held vertically
Specifications
Bracket | |
Stainless Steel | |
For manual and automatic measurements |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
SI Analytics™ Stainless Steel Brackets