Learn More
BRAND™ Standard Ground Joint Funnel
Half-round body
Brand: BRAND™ 94905
320.82 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
40 mm | |
Stem for NS 14/23 | |
Standard Ground Joint Funnel |
Polypropylene | |
10 |
Product Content Correction
Product Title
BRAND™ Standard Ground Joint Funnel > Diameter (Metric)Top: 40 mm
Spot an opportunity for improvement?Share a Content Correction