Learn More
BRAND™ Standard Ground Joint Funnel
Half-round body
269.00 DKK - 460.00 DKK
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | ||||||
11804120
|
BRAND™
94905 |
40 mm |
331.00 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11814120
|
BRAND™
94910 |
50 mm |
460.00 DKK
Pack of 10 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
11824120
|
BRAND™
94915 |
80 mm |
269.00 DKK
Pack of 5 |
In Stock
Log in to see stock. |
||||||
Description
Specifications
Polypropylene | |
Standard Ground Joint Funnel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ Standard Ground Joint Funnel