Learn More
Thermo Scientific Chemicals Standard Sample Tube, 3.24mm ID
For use with MKII Magnetic Susceptibility Balances
Brand:  Thermo Scientific Chemicals 014349.KJ
2648.64 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15431067
  • 2661.88 DKK / Pack of 5
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • I.D.: 3.24mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 Pack
Standard Sample Tube
Product Title
Thermo Scientific Chemicals Standard Sample Tube, 3.24mm ID > 5pc