Learn More
Sampling Systems™ SteriWare™ Disposable Tweezers
Pick up both small and large items with a special dual action design. Sampling Systems™ SteriWare™ Disposable Tweezers are ideal for picking up items up to 25mm wide.
Brand:  Sampling Systems™ 8025H-115S
 View more versions of this product
Product Code. 15849421
  • 636.00 DKK / Pack of 100
Estimated Shipment: 03-06-2025
Explore more special offers
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No Cross Contamination
  • No Cleaning required
Specifications
Specifications
HDPE
115 mm
Sterile Forceps
Sterile
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number122055 Certificate TypeCertificate of Compliance Date01/05/2025 Catalog Number8025H-115S
Lot Number120950 Certificate TypeCertificate of Compliance Date16/12/2024 Catalog Number8025H-115S
Lot Number119713 Certificate TypeCertificate of Compliance Date18/09/2024 Catalog Number8025H-115S
Lot Number118008 Certificate TypeCertificate of Compliance Date26/03/2024 Catalog Number8025H-115S
Lot Number070766 Certificate TypeCertificate of Compliance Date14/02/2023 Catalog Number8025H-115S
Product Title
Sampling Systems™ SteriWare™ Disposable Tweezers > Product Type: Sterile Forceps