Learn More
Tocris Bioscience™ SU 3327
Selective JNK inhibitor
Brand:  Tocris Bioscience™ 3607/50
6010.53 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18034432
  • 7650.00 DKK / 50mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
SU 3327 is a selective inhibitor of c-Jun N-terminal kinase (JNK) (IC50 = 0.7 μM). Inhibits the protein-protein interaction between JNK and JIP (IC50 = 239 nM). Displays selectivity over p38 MAPK and Akt. Restores insulin sensitivity in a mouse model of type-2 diabetes. Displays antibiotic activity against a range of bacteria including S.aureus, A.baumanni, C.difficile and M.tuberculosis.
Specifications
Specifications
>99%
50 mg
261.3
Halicin
SU 3327
SU 3327
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only