missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SU 3327
Selective JNK inhibitor
1915.00 DKK - 7650.00 DKK
Specifications
Purity | >99% |
---|---|
Gene Alias | Halicin |
Product Type | SU 3327 |
Molecular Weight (g/mol) | 261.3 |
Inhibitors | SU 3327 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18035902
|
Tocris Bioscience™
3607/10 |
10 mg |
1915.00 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18034432
|
Tocris Bioscience™
3607/50 |
50 mg |
7650.00 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Selective inhibitor of c-Jun N-terminal kinase (JNK) (IC50 = 0.7 μM). Inhibits the protein-protein interaction between JNK and JIP (IC50 = 239 nM). Displays selectivity over p38 MAPK and Akt. Restores insulin sensitivity in a mouse model of type-2 diabetes.Specifications
>99% | |
SU 3327 | |
SU 3327 |
Halicin | |
261.3 |
For Research Use Only