missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Symmetric Dimethyl Arginine Motif Rabbit anti-Human, Mouse, Rat, Clone: 8I0J1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16078-100UL
This item is not returnable.
View return policy
Description
Symmetric Dimethyl Arginine Motif Monoclonal antibody specifically detects Symmetric Dimethyl Arginine Motif in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Symmetric Dimethyl Arginine Motif | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
8I0J1 | |
Western Blot 1:500 - 1:1000 | |
SDMA, Symmetric Dimethylarginine | |
A synthetic peptide corresponding to a sequence containing methylated R. | |
100 μg | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |