missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Symmetric Dimethyl Arginine Motif Rabbit anti-Human, Mouse, Rat, Clone: 8I0J1, Novus Biologicals™
Rabbit Monoclonal Antibody
1460.01 DKK - 4239.64 DKK
Specifications
Antigen | Symmetric Dimethyl Arginine Motif |
---|---|
Clone | 8I0J1 |
Dilution | Western Blot 1:500 - 1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310722
|
Bio-Techne
NBP3-16078-20UL |
20 μg |
1460.01 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18076925
|
Bio-Techne
NBP3-16078-100UL |
100 μg |
4239.64 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Symmetric Dimethyl Arginine Motif Monoclonal antibody specifically detects Symmetric Dimethyl Arginine Motif in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Symmetric Dimethyl Arginine Motif | |
Western Blot 1:500 - 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
A synthetic peptide corresponding to a sequence containing methylated R. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
8I0J1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
SDMA, Symmetric Dimethylarginine | |
IgG | |
Affinity purified |