missing translation for 'onlineSavingsMsg'
Learn More

Symmetric Dimethyl Arginine Motif Rabbit anti-Human, Mouse, Rat, Clone: 8I0J1, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Bio-Techne NBP3-16078-20UL

 View more versions of this product

Product Code. 18310722

  • 1460.01 DKK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Symmetric Dimethyl Arginine Motif Monoclonal antibody specifically detects Symmetric Dimethyl Arginine Motif in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Symmetric Dimethyl Arginine Motif
Monoclonal
Unconjugated
PBS, 0.05% BSA, 50% glycerol, pH7.3
Rabbit
Affinity purified
RUO
Human, Mouse, Rat
Purified
Western Blot
8I0J1
Western Blot 1:500 - 1:1000
SDMA, Symmetric Dimethylarginine
A synthetic peptide corresponding to a sequence containing methylated R.
20 μg
Primary
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers