Learn More
BICASA Tall Laboratory Cabinet
TALL storage cabinet is made from fire-retardant particleboard and has 4 shelves.
5530.00 DKK
Specifications
Cabinet Type | Storage |
---|---|
Certifications/Compliance | EN 16121:2017 Certified |
Dimensions (L x W x H) | (480 + 20) x 600 x 2030 mm |
Door Type | Hinged |
Finish | Powder Coated |
Product Code | Brand | Hinge Side | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Hinge Side | Color | Price | Quantity & Availability | |||||
30057545
|
BICASA
TCD.0600.2003.LH |
Left | Light Gray |
5530.00 DKK
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
30057596
|
BICASA
TCD.0600.2003.LH-A |
Left | Anthracite |
5530.00 DKK
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
30057555
|
BICASA
TCD.0600.2003.LH-B |
Left | Blue |
5530.00 DKK
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
30057606
|
BICASA
TCD.0600.2003.LH-R |
Left | Red |
5530.00 DKK
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
30057593
|
BICASA
TCD.0600.2003-A |
Right | Anthracite |
5530.00 DKK
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
30057576
|
BICASA
TCD.0600.2003-B |
Right | Blue |
5530.00 DKK
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
30057541
|
BICASA
TCD.0600.2003-R |
Right | Red |
5530.00 DKK
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
Description
- Tall Storage Cabinet
- EN 16121:2017 certified / complied
- Metal frame levelling plinth
- Light Grey matt bonderized Polyurethane powder coating
- Fire-retardant particleboard BODY mm 19 thickness
- Light Grey color (2 decorative)
- Steel sheet shelves (height-adjustable)
- Light Grey matt bonderized Polyurethane powder coating
- Solid hinged door
- 270° opening Automatic hold-closing
- Ergonomic anodized and painted aluminum handle
- Wardrobe lock with bar as standard
Specifications
Storage | |
(480 + 20) x 600 x 2030 mm | |
Powder Coated | |
1 | |
4 | |
23.6 in. | |
No | |
Melamine |
EN 16121:2017 Certified | |
Hinged | |
Melamine | |
None | |
None | |
600 mm | |
Storage Unit |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BICASA Tall Laboratory Cabinet