Learn More
Applied Biosystems™ TaqMan™ Array Human CYP450 & other Oxygenases
TaqMan™ Array Human CYP450 and other Oxygenases 96-well Plate contains 92 assays to CYP450 and associated genes, and 4 assays to candidate endogenous control genes.
Brand:  Applied Biosystems™ 4414085
Product Code. 10498264
  • 1812.00 DKK / 1 set
Explore more special offers
Description
Description
The panel of assays in this plate targets genes from 56 CYP450 family. Additionally, there are assays against genes which participate in the CYP450 containing systems, including the cytochrome oxidoreductase and the cytochrome b5 reductases. Also included are the functionally related members of the monooxygenase, monoamine oxygenase and lipoxygenase families. Gene Signature Plates are 96-well plates that are pre-configured with the most appropriate TaqMan Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment.
TaqMan Array Plates contain the same high quality TaqMan Gene Expression Assays that have been available for years in a convenient, dried, 96-well format. Extensive tests have found no difference in performance between TaqMan Assays supplied in the traditional liquid format and assays dried into a TaqMan Array Plate.
Plate map ensures proper data labeling and easy re-ordering. Each pack of TaqMan Array Plates is shipped with a CD containing the plate configuration map along with the details of each TaqMan Assay. This information is imported into the real-time PCR instrument to enable proper labeling of experimental data and easy access to Assay ID numbers for simple reordering.
  • Affordable; minimum order of three plates; ideal for small, medium or large-sized projects
  • Fast delivery; on your bench in days
  • Convenient; dried PCR primer and TaqMan™ probe sets supplied in a 96- well plate; just add master mix and your sample
  • TaqMan Array Plates have been optimized for use with TaqMan Gene Expression Master Mix, but are also compatible with TaqMan Universal PCR Master Mix
  • For studies using cultured cells, TaqMan Array Plates are also compatible (and recommended) with the TaqMan Gene Expression Cells-to-Ct™ Kit
  • TaqMan Array Plates must be run on a real-time PCR instrument with 96-well plate capability
  • The plates have been validated for use on Applied Biosystems™ 7000, 7300, 7500 and 7900HT Fast Real-Time PCR Systems using standard 96-well blocks, standard PCR cycling conditions, and 20μL reaction volumes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C.
96-well plate
MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
TaqMan™
1 standard plate
Human
1 x 20 μL Reactions
Dried Down
Standard
FAM
Standard
Toxicology & Drug Metabolism
Gene Expression
Documents
Manuals
Specification Sheets
Product Title
Applied Biosystems™ TaqMan™ Array Human CYP450 & other Oxygenases > 1 Standard Plate