Learn More
Tenovin-3, MedChemExpress
MedChemExpress Tenovin-3 is a p53 activator.
Brand:  Medchem Express HY-19339
Encompass_Preferred
 View more versions of this product
Product Code. 16471799
  • 725.13 DKK / 1mg
Explore more special offers
Description
Description
Tenovin-3 is a p53 activator.
Specifications
Specifications
Tenovin-3
1011301-27-1
1 mg
C18H21N3OS
DMSO : ≥ 31 mg/mL (94.67 mM)
327.44
99.86%
Solid
White
Research
Cancer-programmed cell death
O=C(NC(NC1=CC=C(N)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2
327.44
Research
Documents
One moment while we fetch your results.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Tenovin-3, MedChemExpress