Learn More
Tenovin-3, MedChemExpress
MedChemExpress Tenovin-3 is a p53 activator.

725.13 DKK - 6757.79 DKK
Specifications
Chemical Name or Material | Tenovin-3 |
---|---|
Physical Form | Solid |
CAS | 1011301-27-1 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16490198
![]() |
Medchem Express
HY-19339 |
5 mg |
1661.76 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16473636
![]() |
Medchem Express
HY-19339 |
25 mg |
4582.41 DKK
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16471799
![]() |
Medchem Express
HY-19339 |
1 mg |
725.13 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16467738
![]() |
Medchem Express
HY-19339 |
10 mg |
2709.17 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16403717
![]() |
Medchem Express
HY-19339 |
50 mg |
6757.79 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tenovin-3 is a p53 activator.Specifications
Tenovin-3 | |
1011301-27-1 | |
Research | |
Cancer-programmed cell death | |
O=C(NC(NC1=CC=C(N)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2 | |
327.44 | |
Research |
Solid | |
White | |
C18H21N3OS | |
DMSO : ≥ 31 mg/mL (94.67 mM) | |
327.44 | |
99.86% |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.