Learn More
MP Biomedicals™ Terrific Broth II, Dehydrated Media
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium
Brand:  MP Biomedicals™ 113046031
 View more versions of this product
Product Code. 11357699
  • 2500.00 DKK / 1kg
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Proprietary formulation
  • Use 50 capsules per liter
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Molecular biology Certified™ FDA
Capsule
Microbiology Media
For growth of Escherichia coli and certified for plasmid and other bacterial cultures in liquid medium
15°C to 30°C
Molecular biology
Yeast extract, Tryptone broth, Sodium Chloride and Vitamins
1000 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Terrific Broth II, Dehydrated Media > 1kg