missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Terrific Broth II, Dehydrated Media
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium
Brand: MP Biomedicals™ 113046041
This item is not returnable.
View return policy
Description
- Proprietary formulation
- Use 50 capsules per liter

Specifications
Molecular biology Certified™ FDA | |
Capsule | |
Microbiology Media | |
For growth of Escherichia coli and certified for plasmid and other bacterial cultures in liquid medium |
15°C to 30°C | |
Molecular biology | |
Yeast extract, Tryptone broth, Sodium Chloride and Vitamins | |
5000 g |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Terrific Broth II, Dehydrated Media > 5kg
Spot an opportunity for improvement?Share a Content Correction