Learn More
Julabo™ Test Tube Racks for Open Heating Bath Circulator
Brand:  Julabo™ 8970347
 View more versions of this product
Product Code. 10387012
  • 1135.00 DKK / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Test tube rack
SW22, SW23, TW12, TW20, TW8 shaking water bath
21 Test Tubes, 30/31 mm dia.
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Julabo™ Test Tube Racks for Open Heating Bath Circulator > Stainless Steel; Holds 21 Test tubes, Dia.: 30/31mm