Learn More
Fisherbrand™ Test Tubes, Soda lime, Without Rim, Medium Wall
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
Brand:  Fisherbrand™ 90101216
377.20 DKK valid until 2025-03-31
Use promo code "24162" to get your promotional price.
 View more versions of this product
Product Code. 17991132
  • 533.00 DKK / Pack of 100
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Manufactured from soda lime glass
  • Thick-walled and therefore mechanically very resistant
  • Available with beaded rim or straight rim
  • With round bottom
TRUSTED_SUSTAINABILITY
Specifications
Specifications
35 mL
Soda Lime Glass
Non-sterile
No
Not Included
0.9 mm
180 mm
Cylindrical
Not Autoclavable
18 mm
Test Tube
Certificates
View accessory products
Special Offers
Special Offers
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24162
    Expires2025-03-31
    Days
    12
    Hours
    22
    Minutes
    10
    Seconds
    28
    New Offer
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
    New Offer
    Save Now
Product Content Correction
Product Title
Fisherbrand™ Test Tubes, Soda lime, Without Rim, Medium Wall > Capacity (Metric): 35 mL