Learn More
Fisherbrand™ Test Tubes, Soda lime, Without Rim, Medium Wall
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
168.16 DKK - 1825.00 DKK
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
15802265
|
Fisherbrand™
90101004 |
3 mL | 75 mm | 10 mm |
168.16 DKK
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
17901132
|
Fisherbrand™
90101223 |
5 mL | 75 mm | 12 mm |
733.00 DKK
Pack of 1000 |
Estimated Shipment: 12-08-2025
Log in to see stock. |
||||
17991142
|
Fisherbrand™
90101224 |
7 mL | 100 mm | 12 mm |
887.00 DKK
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||
17911122
|
Fisherbrand™
90101213 |
10 mL | 100 mm | 14 mm |
1145.00 DKK
Pack of 1000 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
17921122
|
Fisherbrand™
90101214 |
12 mL | 100 mm | 16 mm |
1180.00 DKK
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||
17971142
|
Fisherbrand™
90101225 |
18 mL | 125 mm | 16 mm |
1245.00 DKK
Pack of 1000 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
17951142
|
Fisherbrand™
90101226 |
22 mL | 150 mm | 16 mm |
1410.00 DKK
Pack of 1000 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
17901122
|
Fisherbrand™
90101215 |
23 mL | 160 mm | 16 mm |
1825.00 DKK
Pack of 1000 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
15802275
|
Fisherbrand™
90101022 |
29 mL | 150 mm | 18 mm |
506.00 DKK
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
17991132
|
Fisherbrand™
90101216 |
35 mL | 180 mm | 18 mm |
533.00 DKK
Pack of 100 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||
15812275
|
Fisherbrand™
90101023 |
55 mL | 150 mm | 24 mm |
298.00 DKK
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||
Description
- Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
- Manufactured from soda lime glass
- Thick-walled and therefore mechanically very resistant
- Available with beaded rim or straight rim
- With round bottom
Specifications
Soda Lime Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Test Tubes, Soda lime, Without Rim, Medium Wall