Learn More
tetrahydrothiopyran-4-carbonitrile, 97%, Thermo Scientific™
839.64 DKK - 2541.99 DKK
Chemical Identifiers
CAS | 195503-40-3 |
---|---|
Molecular Formula | C6H9NS |
Molecular Weight (g/mol) | 127.21 |
MDL Number | MFCD09025909 |
InChI Key | DQKVZWAEWJUKAX-UHFFFAOYSA-N |
Synonym | tetrahydro-2h-thiopyran-4-carbonitrile, tetrahydrothiopyran-4-carbonitrile, 2h-thiopyran-4-carbonitrile,tetrahydro, 2h-thiopyran-4-carbonitrile, tetrahydro, 4-cyanothiane, 4-cyanothian-4-yl, tetrahydro-thiopyran-4-carbonitrile |
PubChem CID | 21598641 |
IUPAC Name | thiane-4-carbonitrile |
SMILES | N#CC1CCSCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10012514
|
Thermo Scientific Maybridge
CC61916DA |
1 g | Amber Glass Bottle | N/A | N/A | N/A | |||||
10442684
|
Thermo Scientific Maybridge
CC61916DE |
5 g | Amber Glass Bottle | N/A | N/A | N/A | |||||
Description
Chemical Identifiers
195503-40-3 | |
127.21 | |
DQKVZWAEWJUKAX-UHFFFAOYSA-N | |
21598641 | |
N#CC1CCSCC1 |
C6H9NS | |
MFCD09025909 | |
tetrahydro-2h-thiopyran-4-carbonitrile, tetrahydrothiopyran-4-carbonitrile, 2h-thiopyran-4-carbonitrile,tetrahydro, 2h-thiopyran-4-carbonitrile, tetrahydro, 4-cyanothiane, 4-cyanothian-4-yl, tetrahydro-thiopyran-4-carbonitrile | |
thiane-4-carbonitrile |
Specifications
195503-40-3 | |
C6H9NS | |
tetrahydro-2h-thiopyran-4-carbonitrile, tetrahydrothiopyran-4-carbonitrile, 2h-thiopyran-4-carbonitrile,tetrahydro, 2h-thiopyran-4-carbonitrile, tetrahydro, 4-cyanothiane, 4-cyanothian-4-yl, tetrahydro-thiopyran-4-carbonitrile | |
N#CC1CCSCC1 | |
127.21 | |
127.21 | |
Tetrahydrothiopyran-4-carbonitrile |
97% | |
MFCD09025909 | |
DQKVZWAEWJUKAX-UHFFFAOYSA-N | |
thiane-4-carbonitrile | |
21598641 | |
97% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
tetrahydrothiopyran-4-carbonitrile, 97%, Thermo Scientific™