Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 30 Layer
Apply directly to floor
Brand: Texwipe™ AMA254581B
2313.25 DKK valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
Apply directly to floor
- Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
- Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
45 x 25 in. | |
30 Sheets/Mat | |
Blue |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Days
18Hours
1Minutes
20Seconds
36