Learn More
Eppendorf™ Thermal printer DPU-S445
Thermal line dot printer with high printing speed and clear type face production.
Brand: Eppendorf™ 6135011000
6104.32 DKK valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Automatic print-out of results
- Easy and direct connection via USB
- Compact and lightweight
- Easy paper orientation
- Compatible with the BioPhotometer D30 and the BioSpectrometer product line
Specifications
BioPhotometer D30 and the BioSpectrometer product line | |
Thermal Dot Printer | |
Max printing speed: 90 mm/sec, Dots/line: 832, Resolution: 8 dots/mm | |
EU Plug |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
5Hours
0Minutes
5Seconds
5
Product Content Correction
Product Title
Eppendorf™ Thermal printer DPU-S445 > EU Plug
Spot an opportunity for improvement?Share a Content Correction