Learn More
Eppendorf™ Thermal printer DPU-S445
Thermal line dot printer with high printing speed and clear type face production.
6425.60 DKK
Specifications
For Use With (Equipment) | BioPhotometer D30 and the BioSpectrometer product line |
---|---|
Type | Thermal Dot Printer |
Description | Max printing speed: 90 mm/sec, Dots/line: 832, Resolution: 8 dots/mm |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
15723275
|
Eppendorf™
6135011000 |
EU Plug |
6425.60 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Automatic print-out of results
- Easy and direct connection via USB
- Compact and lightweight
- Easy paper orientation
- Compatible with the BioPhotometer D30 and the BioSpectrometer product line
Specifications
BioPhotometer D30 and the BioSpectrometer product line | |
Thermal Dot Printer | |
Max printing speed: 90 mm/sec, Dots/line: 832, Resolution: 8 dots/mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Eppendorf™ Thermal printer DPU-S445