Learn More
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck
Shop All Dupont Products
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
3X-Large
Description
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash
- Breathable and exceptionally low linting
Specifications
Specifications
Product Type | Controlled Environments Lab Coat |
Size | 3X-Large |
Material | Polyethylene |
Color | White |
Closure Type | Snaps |
Sterility | Non-sterile |
Model | IC270B0 |
Seam Type | Bound |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
New Offer Save Up to 92%
Product Content Correction
Product Title
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck >
Spot an opportunity for improvement?Share a Content Correction