Learn More
Valbenazine, MedChemExpress
MedChemExpress Valbenazine (NBI-98854) is a vesicular monoamine transporter 2 (VMAT2) inhibitor with the Ki of 110-190 nM.
Brand: Medchem Express HY-16771
Description
Valbenazine (NBI-98854) is a vesicular monoamine transporter 2 (VMAT2) inhibitor with the Ki of 110-190 nM.Specifications
Valbenazine | |
Light Yellow | |
C24H38N2O4 | |
NBI-98854 | |
O=C([C@@H](N)C(C)C)O[C@@H]1C[C@]2([H])C3=CC(OC)=C(OC)C=C3CCN2C[C@H]1CC(C)C | |
418.57 | |
Research | |
Neuroscience-Neuromodulation |
1025504-45-3 | |
Research | |
10 mg | |
DMSO : 50 mg/mL (119.45 mM; Need ultrasonic) | |
418.57 | |
98.0% | |
Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only