Learn More
Kartell™ Variable Volume Pluripet II Micropipettor
Ensure a high degree of precision and accuracy combined with a lower cost. Kartell™ Variable Volume Pluripet II Micropipettor is useful for applications in Molecular Biology, Microbiology, Immunology, Analytical Chemistry, etc.
1525.00 DKK - 1580.00 DKK
Specifications
Certifications/Compliance | EC/IVD, ISO8655 |
---|---|
Autoclavable | Autoclavable |
Type | Micropipette |
Product Code | Brand | Volume (Metric) | Compatible Tips | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Compatible Tips | Price | Quantity & Availability | ||||||
10650954
|
Kartell™
1296000 |
0.2 to 2 μL | Kartell™ 956 |
1565.00 DKK
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10533045
|
Kartell™
1300000 |
2 to 20 μL | Kartell™ 959 | N/A | N/A | N/A | |||||
10213763
|
Kartell™
1301000 |
20 to 200 μL | Kartell™ 960 |
1580.00 DKK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
10745384
|
Kartell™
1321000 |
20 to 100 μL | Kartell™ 960 | N/A | N/A | N/A | |||||
10571125
|
Kartell™
1325000 |
1 to 5 mL | Kartell™ 984 |
1580.00 DKK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
10671144
|
Kartell™
1329000 |
2 to 10 mL | Kartell™ 943 | N/A | N/A | N/A | |||||
Descripción
- Made of high quality plastic and equipped with steel piston for prolonged use over time
- The handle connection ensures a more comfortable and non-fatiguing grip
- The desired volume can be set by turning the emptying button
- It proves easy and safe thanks to the snap selection
- The large display ensures simple reading of the set volume value
- The button of each micropipettor has a specific colour so identification of the usable volume interval is simple
- The tip holder cone can house a wide variety of tips
- The recalibration device ensures quick and precise recalibrations
- The robust structure of the body ensures high mechanical and chemical resistances
Especificaciones
EC/IVD, ISO8655 | |
Autoclavable | |
Micropipette |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Kartell™ Variable Volume Pluripet II Micropipettor