Learn More
VD2-d3, MedChemExpress
MedChemExpress VD2-d3 is the deuterium labeled vitamin D2.
Brand: Medchem Express HY-15330
This item is not returnable.
View return policy
Description
VD2-d3 is the deuterium labeled vitamin D2.Specifications
VD2-d3 | |
1 mg | |
Research | |
DMSO : 20 mg/mL (50.04 mM; ultrasonic and warming and heat to 80°C) | |
399.67 | |
95.46% | |
Metabolism-sugar/lipid metabolism |
1217448-46-8 | |
White | |
C28H41D3O | |
O[C@@H](CCC/1=C([2H])\[2H])CC1=C([2H])\C=C2[C@@](CC[C@@H]3[C@@H](/C=C/[C@H](C)C(C)C)C)([H])[C@]3(C)CCC\2 | |
399.67 | |
Research | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.