Learn More
Tocris Bioscience™ VU 0546110
Selective SLO3 potassium channel inhibitor
Brand: Tocris Bioscience™ 7956/10
863.20 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
VU 0546110 is a selective inhibitor of the sperm-specific potassium channel SLO3 (IC50 = 64 μM). It shows 40-fold selectivity over SLO1. It completely blocks heterologous SLO3 currents and endogenous K+ currents. VU 0546110 inhibits sperm hyperpolarisation and prevents its hyperactivated mobility.Specifications
1-[4-[[(4-Methoxyphenyl)thio]methyl]-1-piperidinyl]-2-[4-[(1-methylethyl)sulfonyl]phenyl]ethanone | |
10 mg | |
C24H31NO4S2 |
1421523-00-3 | |
Non-selective/Other Potassium Channel Blockers | |
0.98 |