missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 0546110

Selective SLO3 potassium channel inhibitor

1055.00 DKK - 3570.00 DKK

Specifications

Chemical Name or Material 1-[4-[[(4-Methoxyphenyl)thio]methyl]-1-piperidinyl]-2-[4-[(1-methylethyl)sulfonyl]phenyl]ethanone
CAS 1421523-00-3
Target Non-selective/Other Potassium Channel Blockers
Molecular Formula C24H31NO4S2
Purity 0.98
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18034272
View Documents
Tocris Bioscience™
7956/50
50 mg
3570.00 DKK
50mg
Please to purchase this item. Need a web account? Register with us today!
18015852
View Documents Promotion Details
Tocris Bioscience™
7956/10
10 mg
1055.00 DKK
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VU 0546110 is a selective inhibitor of the sperm-specific potassium channel SLO3 (IC50 = 64 μM). It shows 40-fold selectivity over SLO1. It completely blocks heterologous SLO3 currents and endogenous K+ currents. VU 0546110 inhibits sperm hyperpolarisation and prevents its hyperactivated mobility.
Specifications

Specifications

1-[4-[[(4-Methoxyphenyl)thio]methyl]-1-piperidinyl]-2-[4-[(1-methylethyl)sulfonyl]phenyl]ethanone
Non-selective/Other Potassium Channel Blockers
0.98
1421523-00-3
C24H31NO4S2
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications