missing translation for 'onlineSavingsMsg'
Learn More

BICASA Wall Bench MECC Series with Sink

Wall Bench MECC Series with Sink, complete with lateral pedestals, MECC service spine, C-Frame side support, cross-bar, PP sink countertop with 1 basin, water mixer, eyewash and pegboard included.

Brand:  BICASA MS.1200.900.2100.PP-B

 View more versions of this product

Product Code. 30057544

  • 35445.00 DKK / Each
Estimated Shipment: 04-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Laboratory benches MECC Series
  • PEDESTALS: Aluminum drawn profile (150 x 25 mm and 150 x 50 mm) with vertical slots
  • SERVICE SPINE: Modular steel frame assembled to the pedestals at different heights with utility panels horizontal rows
  • Trunking structure with passageway for piping and wirings
  • UTILITY STRIP: predrilled metal band at countertop height
  • UTILITY PANELS: Moulded Polypropylene PP resin cover (V0 flame retarded - IEC 60695-11-10 / -20)
  • C-FRAME WORKSTATIONS: certified according to EN 13150:2001 and BRG 234/09.06
  • C-shaped steel tubular frame levelling supports - Horizontal cross-bars - Lateral metal sheeting
  • COUNTERTOPS: Independent units - installed and fixed above the C-FRAME
  • COLORS: METAL: Light Grey matt bonderized Polyurethane powder coating
  • UTILITY MECC PANELS: Silver Grey (frontal plate) - ABS Electric panel rear box: Black
  • FIRE-RETARDANT PARTICLEBOARD 19 mm panels: Light Grey (2 decorative)
  • TAPS: On turret - Epoxy powder coating - Light Grey color
  • Locking on utility strip or on countertop with threaded inlet feed sleeve and lock nut
  • Fluid identification (handle according to EN13792:2000)
  • REAGENT HOLDER SHELF: coated metal - cantilever
  • PEGBOARD: Plastic (acrylic stand) panel
Specifications

Specifications

Wall Bench
Blue
1200 x 900 x 2100 mm
Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BICASA Wall Bench MECC Series with Sink > Dimensions (L x W x H): 1200 x 900 x 2100 mm, Color Facade: Blue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.