missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BICASA Wall Bench MECC Series with Sink
Wall Bench MECC Series with Sink, complete with lateral pedestals, MECC service spine, C-Frame side support, cross-bar, PP sink countertop with 1 basin, water mixer, eyewash and pegboard included.
19925.00 DKK - 35445.00 DKK
Product Code | Brand | Dimensions (L x W x H) | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Color | Price | Quantity & Availability | |||||
30057591
|
BICASA
MS.0600.900.930.PP-A |
600 x 900 x 930 mm | Anthracite |
19925.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057556
|
BICASA
MS.0600.900.930.PP-B |
600 x 900 x 930 mm | Blue |
19925.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057608
|
BICASA
MS.0600.900.930.PP-R |
600 x 900 x 930 mm | Red |
19925.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057539
|
BICASA
MS.0600.900.2100.PP-A |
600 x 900 x 2100 mm | Anthracite |
26690.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057590
|
BICASA
MS.0600.900.2100.PP-B |
600 x 900 x 2100 mm | Blue |
26690.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057548
|
BICASA
MS.0600.900.2100.PP-R |
600 x 900 x 2100 mm | Red |
26690.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057573
|
BICASA
MS.1200.900.930.PP-A |
1200 x 900 x 930 mm | Anthracite |
27235.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057550
|
BICASA
MS.1200.900.930.PP-B |
1200 x 900 x 930 mm | Blue |
27235.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057574
|
BICASA
MS.1200.900.930.PP-R |
1200 x 900 x 930 mm | Red |
27235.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057559
|
BICASA
MS.1200.900.2100.PP-A |
1200 x 900 x 2100 mm | Anthracite |
35445.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057544
|
BICASA
MS.1200.900.2100.PP-B |
1200 x 900 x 2100 mm | Blue |
35445.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
30057586
|
BICASA
MS.1200.900.2100.PP-R |
1200 x 900 x 2100 mm | Red |
35445.00 DKK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
Description
- Laboratory benches MECC Series
- PEDESTALS: Aluminum drawn profile (150 x 25 mm and 150 x 50 mm) with vertical slots
- SERVICE SPINE: Modular steel frame assembled to the pedestals at different heights with utility panels horizontal rows
- Trunking structure with passageway for piping and wirings
- UTILITY STRIP: predrilled metal band at countertop height
- UTILITY PANELS: Moulded Polypropylene PP resin cover (V0 flame retarded - IEC 60695-11-10 / -20)
- C-FRAME WORKSTATIONS: certified according to EN 13150:2001 and BRG 234/09.06
- C-shaped steel tubular frame levelling supports - Horizontal cross-bars - Lateral metal sheeting
- COUNTERTOPS: Independent units - installed and fixed above the C-FRAME
- COLORS: METAL: Light Grey matt bonderized Polyurethane powder coating
- UTILITY MECC PANELS: Silver Grey (frontal plate) - ABS Electric panel rear box: Black
- FIRE-RETARDANT PARTICLEBOARD 19 mm panels: Light Grey (2 decorative)
- TAPS: On turret - Epoxy powder coating - Light Grey color
- Locking on utility strip or on countertop with threaded inlet feed sleeve and lock nut
- Fluid identification (handle according to EN13792:2000)
- REAGENT HOLDER SHELF: coated metal - cantilever
- PEGBOARD: Plastic (acrylic stand) panel
Specifications
Wall Bench | |
Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BICASA Wall Bench MECC Series with Sink