Learn More
Water, Certified AR for Analysis, Certified AR, Fisher Chemical™
Brand: Fisher Chemical W/0100/24
644.72 DKK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Water, Certified AR for Analysis, Certified AR
Specifications
Bag in box | |
10 L |
Safety and Handling
- Water
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2527027 | Certificate of Analysis | 13/02/2025 | W/0100/FP21,W/0100/24,W/0100/25,W/0100/27,W/0100/21 |
2527026 | Certificate of Analysis | 13/02/2025 | W/0100/FP21,W/0100/24,W/0100/25,W/0100/27,W/0100/21 |
2525943 | Certificate of Analysis | 06/02/2025 | W/0100/FP21,W/0100/24,W/0100/25,W/0100/27,W/0100/21 |
2525942 | Certificate of Analysis | 06/02/2025 | W/0100/FP21,W/0100/24,W/0100/25,W/0100/27,W/0100/21 |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
4Hours
7Minutes
29Seconds
28 -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
4Hours
7Minutes
29Seconds
28 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
35Hours
7Minutes
29Seconds
28New Offer Save Up to 70%