Learn More
Water, Certified AR for Analysis, Certified AR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
5 L
10 L
25 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Unit Size:
10 L
25 L
5 L
Description
Water, Certified AR for Analysis, Certified AR
Specifications
Specifications
Packaging | HDPE plastic bottle |
Quantity | 5 L |
Documents
Scientific Resources
Brochures
Fisherbrand Focus on pH and electrochemistry
File size: 9.1 MB
Fisherbrand Focus on pH and electrochemistry
File size: 2.5 MB
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
2544983 | Certificate of Analysis | 17/07/2025 | W/0100/FP21,W/0100/24,W/0100/25,W/0100/27,W/0100/21 |
2544984 | Certificate of Analysis | 17/07/2025 | W/0100/FP21,W/0100/24,W/0100/25,W/0100/27,W/0100/21 |
2543367 | Certificate of Analysis | 03/07/2025 | W/0100/FP21,W/0100/24,W/0100/25,W/0100/27,W/0100/21 |
2543366 | Certificate of Analysis | 03/07/2025 | W/0100/FP21,W/0100/24,W/0100/25,W/0100/27,W/0100/21 |
2541322 | Certificate of Analysis | 13/06/2025 | W/0100/FP21,W/0100/24,W/0100/25,W/0100/27,W/0100/21 |
Safety and Handling
Product Identifier
- Water
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
-
Multiple Manufacturers
Chemicals Monthly Promotion
Take advantage of our exclusive monthly promotions on high-quality chemicals.Promo Code: 24202Expires: 2025-07-31
New Offer Save Up to 74%
Product Content Correction
Product Title
Water, Certified AR for Analysis, Certified AR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction