Learn More
Cytiva Whatman 603T Extraction Thimbles
Brand:  Cytiva 10350416
Product Code. 12414342
  • 387.00 DKK / Pack of 25
Estimated Shipment: 21-08-2025
to see stock.
Explore more special offers
Specifications
1 Cs.
50 mm
Extraction Thimbles
Product Title
Cytiva Whatman 603T Extraction Thimbles > Size: 27 x 25 x 50mm