Learn More
uvex X-Tended Support BOA™ Black/Yellow 65682 Low Shoe - Width 11
Uvex 1 safety shoes protect and support the natural movement of the human body by reducing pressure and strain; protection classes S1 P and S3
1150.00 DKK
Specifications
Color | Black, Yellow |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17262291
|
uvex
6568235 |
35 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17272291
|
uvex
6568236 |
36 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17282291
|
uvex
6568237 |
37 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17292291
|
uvex
6568238 |
38 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17202301
|
uvex
6568239 |
39 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17212301
|
uvex
6568240 |
40 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17222301
|
uvex
6568241 |
41 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17232301
|
uvex
6568242 |
42 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17242301
|
uvex
6568243 |
43 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17252301
|
uvex
6568244 |
44 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17262301
|
uvex
6568245 |
45 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17272301
|
uvex
6568246 |
46 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17282301
|
uvex
6568247 |
47 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
17292301
|
uvex
6568248 |
48 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
|||||
17202311
|
uvex
6568249 |
49 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
|||||
17212311
|
uvex
6568250 |
50 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
|||||
17222311
|
uvex
6568251 |
51 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
|||||
17232311
|
uvex
6568252 |
52 |
1150.00 DKK
1 pair |
Estimated Shipment: 02-07-2025
Log in to see stock. |
Vous avez déjà un compte ? Connectez-vous pour voir vos prix et disponibilités. Nouveau ici ? Inscrivez-vous dès aujourd'hui ! |
|||||
Description
Uvex 1 X-Tended Support Shoe 6568.2 with BOA™ Fit System S1 P SRC
- Sporty, multifunctional safety shoe
- Lightweight and flexible
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Penetration-resistant, non-metallic midsole in accordance with the latest safety standards, does not restrict flexibility
- Made with new developed technology for extended wearing comfort
- Uvex Climazone technology for optimum wearer comfort
- Breathable, perforated, high-tech microvelour upper material
- Distance mesh lining provides optimum breathability
- Made from synthetic materials, suitable for people allergic to chrome
- BOA™ Fit System Dial for custom comfort that's consistent all day
- Virtually seamless design to eliminate pressure points
- Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and at front of foot
- Soft, padded collar and tongue
- Medial and lateral heel support for foot through especially shaped side frame to protect against ankle injuries, for guidance and impact protection
- Outstanding dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
- Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Fulfills ESD requirements: resistance to ground Kleiner 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6568.2
- Standard EN ISO 20345:2011 S1 P SRC
- Sizes: 35 to 52
Spécification
Black, Yellow | |
Dual-density polyurethane (PU/PU) | |
Reusable | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Standard Width | |
Yes | |
S1 | |
ESD Safety Shoe |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.