Learn More
YK-4-279, MedChemExpress
MedChemExpress YK 4-279 is an inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1.
Brand:  Medchem Express HY-14507
Encompass_Preferred
 View more versions of this product
Product Code. 16406088
  • 1107.84 DKK / 10mg
Explore more special offers
Description
Description
YK 4-279 is an inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1.
Specifications
Specifications
YK-4-279
Off-White
C17H13Cl2NO4
DMSO : 25 mg/mL (68.27 mM; Need ultrasonic)
366.2
99.61%
Solid
1037184-44-3
Research
10 mg
ClC1=C2C(C(O)(CC(C3=CC=C(OC)C=C3)=O)C(N2)=O)=C(Cl)C=C1
366.2
Research
Cancer-programmed cell death
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
YK-4-279, MedChemExpress