missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YY1 Rabbit anti-Human, Mouse, Rat, Clone: S02-8A3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14987-25UL
This item is not returnable.
View return policy
Description
YY1 Monoclonal antibody specifically detects YY1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
YY1 | |
Monoclonal | |
Unconjugated | |
DELTA, Delta transcription factor, INO80SYY-1, NF-E1INO80 complex subunit S, transcriptional repressor protein YY1, UCRBP, Yin and yang 1, Yin and Yang 1 protein, YIN-YANG-1, YY1 transcription factor | |
A synthetic peptide of human YY1 (Uniprot#: P25490) | |
25 μg | |
Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
7528 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
S02-8A3 | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |