missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YY1 Rabbit anti-Human, Mouse, Rat, Clone: S02-8A3, Novus Biologicals™
Rabbit Monoclonal Antibody
2180.29 DKK - 4407.01 DKK
Specifications
Antigen | YY1 |
---|---|
Clone | S02-8A3 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18301435
|
Novus Biologicals
NBP3-14987-25UL |
25 μg |
2180.29 DKK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18310782
|
Novus Biologicals
NBP3-14987-100UL |
100 μg |
4407.01 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
YY1 Monoclonal antibody specifically detects YY1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
YY1 | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
7528 | |
IgG | |
Affinity-purified |
S02-8A3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DELTA, Delta transcription factor, INO80SYY-1, NF-E1INO80 complex subunit S, transcriptional repressor protein YY1, UCRBP, Yin and yang 1, Yin and Yang 1 protein, YIN-YANG-1, YY1 transcription factor | |
A synthetic peptide of human YY1 (Uniprot#: P25490) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |